Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-KIZUNA (Thr379) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564682 Supplier Invitrogen™ Supplier No. PA564682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-KIZUNA (Thr379) Polyclonal Antibody detects endogenous levels of KIZUNA only when phosphorylated at Thr379.

Centrosomal protein required for establishing a robust mitotic centrosome architecture that can endure the forces that converge on the centrosomes during spindle formation. Required for stabilizing the expanded pericentriolar material around the centriole.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-KIZUNA (Thr379)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIZ
Gene Accession No. Q2M2Z5
Gene Alias C20orf19; Centrosomal protein kizuna; Gm114; HT013; KIZ; Kizuna; kizuna centrosomal protein; NCRNA00153; PLK1S1; polo-like kinase 1 substrate 1; RGD1307071; RP69
Gene Symbols KIZ
Host Species Rabbit
Immunogen A synthesized peptide derived from human Kizuna (Phospho-Thr379).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55857
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.