Learn More
Invitrogen™ Phospho-KV3.1 (Ser503) Polyclonal Antibody

Description
This antibody is specific for the ~100 kDa Voltage Gated Potassium Channel KV3 1-subunit protein phosphorylated at Ser503 in Western blots of rat brain extracts.
Specifications
Specifications
| Antigen | Phospho-KV3.1 (Ser503) |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | KCNC1 |
| Gene Accession No. | P15388, P25122 |
| Gene Alias | C230009H10Rik; EPM7; FLJ41162; FLJ42249; FLJ43491; KCNC1; Kcr2-1; Krc2 1; KShIIIB; KV3.1; KV4; MGC129855; NGK2; NGK2-KV4; Potassium channel gene 1 (alternative splicing product described in Luneau et al 1991); potassium channel, voltage gated Shaw related subfamily C, member 1; potassium channel, voltage gated Shaw-related subfamily C, member 1; potassium voltage gated channel, Shaw-related subfamily, member 1; potassium voltage-gated channel subfamily C member 1; potassium voltage-gated channel, Shaw-related subfamily, member 1; RAW2; Shaw; Shaw-related voltage-gated potassium channel protein 1; voltage-gated potassium channel KCNC1; voltage-gated potassium channel protein KV3.1; voltage-gated potassium channel subunit Kv3.1; Voltage-gated potassium channel subunit Kv4 |
| Gene Symbols | KCNC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.