Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-KV4.2 (Ser616) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564525 Supplier Invitrogen™ Supplier No. PA564525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-KCND2 (Ser616) Polyclonal Antibody detects endogenous levels of KCND2 only when phosphorylated at Ser616.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes-shaker, shaw, shab, and shal-have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shal-related subfamily, members of which form voltage-activated A-type potassium ion channels and are prominent in the repolarization phase of the action potential. This member mediates a rapidly inactivating, A-type outward potassium current which is not under the control of the N terminus as it is in Shaker channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-KV4.2 (Ser616)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCND2
Gene Accession No. Q63881, Q9NZV8, Q9Z0V2
Gene Alias AI839615; AW555701; KCND2; KIAA1044; KV4.2; mKIAA1044; MNCb-7013; potassium channel alpha subunit Kv4.2; potassium channel Kv4.2; potassium channel, voltage gated Shal related subfamily D, member 2; potassium channel, voltage-gated Shal-related subfamily D, member 2; potassium voltage gated channel, Shal-related family, member 2; potassium voltage-gated channel subfamily D member 2; potassium voltage-gated channel, Shal-related family, member 2; potassium voltage-gated channel, Shal-related subfamily, member 2; R75121; RK5; Shal1; voltage-gated potassium channel; voltage-gated potassium channel subunit Kv4.2; voltage-sensitive potassium channel
Gene Symbols KCND2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Kv4.2/KCND2 (Phospho-Ser616).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16508, 3751, 65180
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.