Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho Lamin A/C (Ser22) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABT138950UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT1389-50UL 50 μL
ABT1389-200 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABT1389-50UL Supplier MilliporeSigma Supplier No. ABT138950UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Phospho Lamin A/C (Ser22).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho Lamin A/C (Ser22)
Applications Western Blot, Inhibition Assays
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LMNA,LMN1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 11 amino acids from human Lamin A/C surrounding phosphorylated Serine 22.
Purification Method Affinity Purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_733821
Test Specificity This rabbit polyclonal antibody detects human Lamin A/C phosphorylated on Serine 22.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.