Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Lamin A/C (Ser22) Recombinant Rabbit Monoclonal Antibody (LaminACS22-CF12), FITC, Invitrogen™

Catalog No. PIMA537431
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537431 10 Tests
PIMA537183 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537431 Supplier Invitrogen™ Supplier No. MA537431
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Lamin A/C (Ser22)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone LaminACS22-CF12
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene LMNA
Gene Accession No. P02545, P48678
Gene Alias 70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg
Gene Symbols LMNA
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser22 of human phospho Lamin A/C.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16905, 4000
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.