Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Lamin A/C (Ser390) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | Phospho-Lamin A/C (Ser390) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Inhibition Assays |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABT1388-50UL
|
MilliporeSigma
ABT138850UL |
50 μL |
Each for $192.35
|
|
|||||
ABT1388-200
|
MilliporeSigma
ABT1388200UL |
200 μL |
Each for $452.71
|
|
|||||
Description
Anti-Phospho-Lamin A/C (Ser390) is a rabbit polyclonal antibody that detects Lamin A/C phosphorylated on Serine 390 and has been tested for use in Western Blotting and Peptide Inhibition Assay.Specifications
Phospho-Lamin A/C (Ser390) | |
Western Blot, Inhibition Assays | |
Unconjugated | |
Rabbit | |
Cell Structure | |
NP_733821 | |
KLH-conjugated linear peptide corresponding to 11 amino acids from human Lamin A/C surrounding phosphorylated Serine 390. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
LMNA,LMN1 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody detects human Lamin A/C phosphorylated on serine 390. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title