Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Lamin A/C (Ser404) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABT1387200 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT1387-200 200 μL
ABT1387-50UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABT1387-200 Supplier MilliporeSigma Supplier No. ABT1387200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Phospho-Lamin A/C (Ser404) is a rabbit polyclonal antibody that detects Lamin A/C phosphorylated on Serine 404 and has been tested for use in Immunofluorescence, Western Blotting, and Peptide Inhibition Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Lamin A/C (Ser404)
Applications Immunofluorescence, Western Blot, Inhibition Assays
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LMNA,LMN1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids from human Lamin A/C surrounding phosphorylated Serine 404.
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_733821
Test Specificity This rabbit polyclonal antibody detects human Lamin A/C phosphorylated on Serine 404.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.