Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-LATS1/LATS2 (Ser909, Ser872) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564591 Supplier Invitrogen™ Supplier No. PA564591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-LATS1/LATS2 (Ser909, Ser872) Polyclonal Antibody detects endogenous levels of LATS1/LATS2 only when phosphorylated at Ser909, Ser872.

The protein encoded by this gene is a putative serine/threonine kinase that localizes to the mitotic apparatus and complexes with cell cycle controller CDC2 kinase in early mitosis. The protein is phosphorylated in a cell-cycle dependent manner, with late prophase phosphorylation remaining through metaphase. The N-terminal region of the protein binds CDC2 to form a complex showing reduced H1 histone kinase activity, indicating a role as a negative regulator of CDC2/cyclin A. In addition, the C-terminal kinase domain binds to its own N-terminal region, suggesting potential negative regulation through interference with complex formation via intramolecular binding. Biochemical and genetic data suggest a role as a tumor suppressor. This is supported by studies in knockout mice showing development of soft-tissue sarcomas, ovarian stromal cell tumors and a high sensitivity to carcinogenic treatments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-LATS1/LATS2 (Ser909, Ser872)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene Lats2
Gene Accession No. O95835, Q7TSJ6, Q8BYR2, Q9NRM7
Gene Alias 4932411G09Rik; AV277261; AW208599; AW228608; h-warts; kinase phosphorylated during mitosis protein; KPM; large tumor suppressor; large tumor suppressor 2; large tumor suppressor homolog 1; Large tumor suppressor homolog 2; large tumor suppressor kinase 1; large tumor suppressor kinase 2; large tumor suppressor, homolog 2; large tumor supressor, homolog 1; LATS (large tumor suppressor, Drosophila) homolog 1; LATS (large tumor suppressor, Drosophila) homolog 2; LATS, large tumor suppressor, homolog 1; LATS, large tumor suppressor, homolog 2; lats1; Lats2; RGD1564085; serine/threonine kinase KPM; serine/threonine-protein kinase kpm; Serine/threonine-protein kinase LATS1; serine/threonine-protein kinase LATS2; WARTS; WARTS protein kinase; warts-like kinase; wts
Gene Symbols Lats1, Lats2
Host Species Rabbit
Immunogen A synthesized peptide derived from human LATS1/2 (Phospho-Ser909/872).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16798, 26524, 305922, 308265, 50523, 9113
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.