Learn More
Invitrogen™ Phospho-Leptin Receptor (Tyr1141) Polyclonal Antibody

Description
Phospho-Leptin Receptor (Tyr1141) Polyclonal Antibody detects endogenous levels of Leptin Receptor only when phosphorylated at Tyr1141.
Specifications
Specifications
| Antigen | Phospho-Leptin Receptor (Tyr1141) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Lepr |
| Gene Accession No. | P48356, P48357 |
| Gene Alias | B219; CD antigen CD295; CD295; CD295 antigen; db; diabetes; Fa; HuB219; HuB219.2; LEPR; LEP-R; Leprb; LEPRD; LEPROT; leptin receptor; Leptin receptor (fatty); leptin receptor gene-related protein; Modb1; OB receptor; obese-like; obl; Obr; OB-R; OB-RGRP |
| Gene Symbols | Lepr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.