Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-LIMK1/LIMK2 (Ser323, Ser314) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143714 Supplier Invitrogen™ Supplier No. PA5143714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to unphosphorylated LIMK1 (Ser-323) peptide then affinity purified using LIMK1 (Ser-323) peptide (without carrier). The antibody detects a protein with the same mobility as anti-LIMK1 (C-terminus, LP1831) in human A431 cells treated with calyculin A. This band is weak in control cells and is not detected after lambda phosphatase treatment.

Proteins containing LIM motifs are typically involved in cell fate determination and growth control. A family of proteins designated LIM kinases, including LIMK-1 and LIMK-2, has been identified. LIMK-1 has been shown to regulate the stabilization of F-Actin structures and cofilin activity, indicating that LIMK-1 plays a role in a signaling pathway involved in the regulation of cell motility and morphogenesis. LIMK-1 inhibits neuronal differentiation of PC12 cells, and is thought to act by interfering with events downstream of MAPK activation. Expression patterns of LIMK-1 and LIMK-2 suggest that these proteins may have different functions during development. A truncated form of LIMK-2 has been identified in adult testis that is thought to arise from an alternative initiation exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-LIMK1/LIMK2 (Ser323, Ser314)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene LIMK1
Gene Accession No. O54785, P53667, P53668, P53669, P53670, P53671
Gene Alias KIZ-1; LIM domain kinase 1; LIM domain kinase 2; lim kinase 1; LIM kinase 2; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM motif-containing protein kinase 2; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK 2; LIMK1; LIMK-1; LIMK2; LIMK-2; Limk2b; Link2
Gene Symbols LIMK1, LIMK2
Host Species Rabbit
Immunogen LIMK1 (Ser-323) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding serine 323 in human LIMK1. This sequence is conserved in rat and mouse LIMK1, and has high homology to Ser-314 in human LIMK2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16885, 16886, 29524, 3984, 3985, 65172
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.