Learn More
Invitrogen™ Phospho-LIMK1 (Ser310) Polyclonal Antibody

Description
Antibody detects endogenous levels of LIMK1 only when phosphorylated at Ser310.
Specifications
Specifications
| Antigen | Phospho-LIMK1 (Ser310) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | LIMK1 |
| Gene Accession No. | P53667, P53668, P53669 |
| Gene Alias | EC 2.7.11.1; kinase LIMK1; KIZ-1; LIM domain kinase 1; lim kinase 1; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK1; LIMK-1 |
| Gene Symbols | LIMK1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.