Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-LIMK1 (Thr508) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536663
Description
This antibody detects endogenous protein at a molecular weight of 72 kDa. Purity is >95% by SDS-PAGE.
LIMK1 is a serine/threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK1 has also been implicated in brain development. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.
Specifications
| Phospho-LIMK1 (Thr508) | |
| Polyclonal | |
| Unconjugated | |
| LIMK1 | |
| EC 2.7.11.1; kinase LIMK1; KIZ-1; LIM domain kinase 1; lim kinase 1; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK1; LIMK-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16885, 3984, 65172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53667, P53668, P53669 | |
| LIMK1 | |
| Synthetic phosphopeptide derived from human LIMK1 around the phosphorylation site of Threonine 508. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction