Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-LKB1 (Ser428) Polyclonal Antibody
Phospho-LKB1 (Ser428) Polyclonal Antibody
Supplier: Thermo Scientific PA536858
Description
Phospho-LKB1 (Ser428) Polyclonal Antibody for Western Blot, IHC (P)

Specifications
Phospho-LKB1 (Ser428) | |
Polyclonal | |
Unconjugated | |
STK11 | |
AA408040; CAMK group; CAMKL family; hLKB1; liver kinase B1; Liver kinase B1 homolog; LKB 1; LKB1; LKB-1; LKB1(S); Par-4; PJS; polarization-related protein LKB1; R75140; renal carcinoma antigen NY-REN-19; serine/threonine kinase 11; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; serine/threonine-protein kinase STK11; STK11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20869, 314621, 6794 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
D4AE59, Q15831, Q9WTK7 | |
STK11 | |
Synthetic phosphopeptide derived from human LKB1 around the phosphorylation site of Serine 428. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction