Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-LRP-6 (Thr1479) Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | phospho-LRP-6 (Thr1479) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Inhibition Assays |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABS1063-25UG
|
MilliporeSigma
ABS106325UG |
25 μg |
Each for $192.35
|
|
|||||
ABS1063-100
|
MilliporeSigma
ABS1063100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-phospho-LRP-6 (Thr1479), Cat. No. ABS1063, is a rabbit polyclonal antibody that detects Low-density lipoprotein receptor-related protein 6 phosphorylated on Thr 1479 and has been tested for use in Western Blotting and Peptide Inhibition Assay.Specifications
phospho-LRP-6 (Thr1479) | |
Western Blot, Inhibition Assays | |
Unconjugated | |
Rabbit | |
Signaling | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_002327 | |
KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphorylated Threonine 1479 of human Low-density lipoprotein receptor-related protein 6 (LRP6). | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Low-density lipoprotein receptor-related protein 6 | |
LRP6 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects LRP6 phosphorylated on Threonine 1479. | |
∽220 kDa observed; 180.43 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title