Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho LSD1 (Ser112), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1462MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1462MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1462MI Supplier MilliporeSigma™ Supplier No. ABE1462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysine-specific histone demethylase 1A (UniProt Q6ZQ88; also known as BRAF35-HDAC complex protein BHC110, Flavin-containing amine oxidase domain-containing protein 2) is encoded by the Kdm1a (also known as Aof2, Kiaa0601, Lsd1) gene (Gene ID 99982) in murine species. The FAD+-dependent, lysine-specific demethylase (LSD) LSD1 is a component of the REST corepressor (CoREST) complexes that mediate gene repression by catalyzing the demethylation of mono- and di-methylated histone H3 Lys4 (H3K4me1/2). LSD1 is also involved in gene activation process associated with nuclear receptors through demethylation of H3K9me1/2. In addition, LSD1 is known to demethylate non-histone substrates, such as p53 and Dnmt1. LSD1 is reported to be phosphorylated on Ser112 by PKCalpha and thereby mediate PKCalpha signaling-dependent circadian clock regulation. Ser112 phosphorylation induces LSD1 interaction with CLOCK:BMAL1 to facilitate E-box-dependent transcriptional activation. Knockin mice bearing Lsd1(SA/SA) alleles coding for phosphorylation-defective S112A LSD1 mutant are reported to exhibit altered circadian rhythms in locomotor behavior with attenuated rhythmic expression of core clock genes and impaired circadian clock phase resetting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho LSD1 (Ser112)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Kdm1a, Aof2, Kiaa0601, Lsd1
Gene Accession No. Q6ZQ88
Host Species Rabbit
Immunogen Linear peptide corresponding to a mouse LSD1 sequence containing the phosphorylated Ser112.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_598633
Test Specificity The Ser112 designation is based on murine KDM1A/LSD1 sequence (UniProt Q6ZQ88). Murine Ser112 is equivalent to human Ser111 and rat Ser106 (UniProt O60341 & F1MA31). Specificity of this polyclonal antibody was confirmed by dot blot analysis using the immunogen peptide and the corresponding non-phosphorylated peptide, as well as by Western blotting analysis of PMA-induced target phosphorylation, lambda phosphatase treatment of the cell lysate prior to Western blotting completely abolished the target band detection (Nam, H.J., et al. (2014). Mol. Cell 53:791-805).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.