Learn More
Invitrogen™ Phospho-MAP1B (Thr1265) Polyclonal Antibody

Description
Recommended Blocking Buffer for ICC/IF and IHC: 5% (v/v) normal horse serum, 5% (v/v) normal goat serum, 0.2% (w/v) L-lysine, 0.2% (v/v) Triton X-100 in PBS, pH 7.2.
Specifications
Specifications
| Antigen | Phospho-MAP1B (Thr1265) |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.12 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | MAP1B |
| Gene Accession No. | P14873, P15205, P46821 |
| Gene Alias | A230055D22; AI843217; claustrin; DKFZp686E1099; DKFZp686F1345; FLJ38954; futsch; I79_010564; LC1; MAP1 light chain LC1; MAP1(X); MAP1.2; MAP1B; MAP-1B; MAP1B heavy chain; MAP5; microtubule associated protein 1B; microtubule associated protein 1B; microtubule-associated protein 1B; microtubule-associated protein 1B; Microtubule-associated protein 1B (MAP 1B) (MAP1.2) (MAP1(X)); microtubule-associated protein 5; Mtap1b; Mtap5; Mtap-5; Neuraxin; PPP1R102; protein phosphatase 1, regulatory subunit 102 |
| Gene Symbols | MAP1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.