Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MAP1LC3A (Ser12) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986613
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6613 Supplier Bioss Supplier No. BS5507R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Phospho-MAP1LC3A (Ser12)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAP1LC3A
Gene Accession No. Q9H492
Gene Alias 1010001H21Rik; 4922501H04Rik; ATG8E; Autophagy-related protein LC3 A; Autophagy-related ubiquitin-like modifier LC3 A; BOS_13394; LC3; LC3A; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; MAP1ALC3; MAP1BLC3; MAP1LC3A; microtubule associated protein 1 light chain 3 alpha; Microtubule-asso; Microtubule-associated protein 1 light chain 3 alpha; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; Microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; zgc:77094
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MAP1LC3A around the phosphorylation site of Ser12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84557
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.