Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MAP2K4 (Ser257) Recombinant Rabbit Monoclonal Antibody (SEK1MKK4S257-C5), FITC, Invitrogen™
GREENER_CHOICE

Catalog No. PIMA537139
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537139 100 Tests
PIMA537385 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537139 Supplier Invitrogen™ Supplier No. MA537139
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MAP2K4 (Ser257)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone SEK1MKK4S257-C5
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MAP2K4
Gene Accession No. P45985, P47809
Gene Alias C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein
Gene Symbols MAP2K4
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser257 of human phospho SEK1/MKK4.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26398, 287398, 6416
Target Species Human, Mouse, Rat, Monkey
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.