Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MAP3K9/Thr312 (Thr312) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501987494
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7494 Supplier Bioss Supplier No. BS6779R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Activates the JUN N-terminal pathway.

Specifications

Antigen Phospho-MAP3K9/Thr312 (Thr312)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAP3K9
Gene Accession No. P80192, Q3U1V8
Gene Alias E130314H24Rik; MAP3K9; MEKK9; mitogen activated protein kinase kinase kinase 9; Mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity); MLK1; PRKE1
Gene Symbols MAP3K9
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MAP3K9 around the phosphorylation site of Thr312.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338372, 4293, 500690
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.