Learn More
Invitrogen™ Phospho-MAPKAPK2 (Thr334) Polyclonal Antibody
Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | Phospho-MAPKAPK2 (Thr334) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | MAPKAPK2 |
| Gene Accession No. | P49137, P49138 |
| Gene Alias | AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1 |
| Gene Symbols | MAPKAPK2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.