Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MAPRE3 (Ser176) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143685 Supplier Invitrogen™ Supplier No. PA5143685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-absorbed to unphospho-EB3 (Ser-176), before affinity purification using phospho-EB3 (Ser-176) peptide (without carrier). The antibody detects a 32 kDa* protein corresponding to the apparent molecular mass of phosphorylated EB3 on SDS-PAGE immunoblots of mouse brain. This reactivity is significantly reduced after lambda phosphatase treatment.

EB3 is a 282 amino acid homologue of EB1, a protein known to bind APC (adenomatous polyposis coli). It has been shown to bind to tubulin in vitro and microtubules in vivo. Northern blotting shows expressions to be mainly in brain, and to a lesser degree in muscle. EB3 interacts with APCL, a homologue of the tumor suppressor APC, and this interaction may be involved in the stabilization or extension of microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MAPRE3 (Ser176)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene MAPRE3
Gene Accession No. Q5XIT1, Q6PER3, Q9UPY8
Gene Alias AI790651; APC binding protein; EB1 protein family member 3; EB2; EB3; EBF3; EBF3-S; End-binding protein 3; Mapre3; microtubule associated protein RP/EB family member 3; microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3
Gene Symbols MAPRE3
Host Species Rabbit
Immunogen Phospho-EB3 (Ser-176) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding serine 176 in human EB3. This peptide sequence is highly conserved in mouse and rat EB3. This site is conserved in EB2 but the neighboring amino acids are not well conserved.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100732, 22924, 298848
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.