Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MARCKS (Ser158) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537631 Supplier Invitrogen™ Supplier No. PA537631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is NIH-3T3 cells; suggested positive control for ICC/IF is Hela cells.

MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MARCKS (Ser158)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MARCKS
Gene Accession No. P26645, P29966, P30009
Gene Alias 80K protein; 80K-L; 80K-L protein; FLJ14368; FLJ90045; KINC; Macs; MARCKS; MARCS; myristoylated alanine rich protein kinase C substrate; myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein; protein kinase C substrate, 80 kDa protein, light chain
Gene Symbols MARCKS
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of serine 158 (R-F-S(p)-F-K) derived from Human MARCKS.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17118, 25603, 4082
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.