Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MARCKS (Ser167, Ser170) Recombinant Rabbit Monoclonal Antibody (MARCKSS167170-C9), FITC, Invitrogen™

Catalog No. PIMA537438
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537438 10 Tests
PIMA537190 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537438 Supplier Invitrogen™ Supplier No. MA537438
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MARCKS (Ser167, Ser170)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone MARCKSS167170-C9
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MARCKS
Gene Accession No. P26645, P29966, P30009
Gene Alias 80K protein; 80K-L; 80K-L protein; FLJ14368; FLJ90045; KINC; Macs; MARCKS; MARCS; myristoylated alanine rich protein kinase C substrate; myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein; protein kinase C substrate, 80 kDa protein, light chain
Gene Symbols MARCKS
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser167/170 of human phospho MARCKS.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17118, 25603, 4082
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.