Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MARCKS (Ser167, Ser170) Recombinant Rabbit Monoclonal Antibody (MARCKSS167170-C9), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$408.50 - $774.00
Specifications
| Antigen | Phospho-MARCKS (Ser167, Ser170) |
|---|---|
| Clone | MARCKSS167170-C9 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA537437
|
Invitrogen™
MA537437 |
20 μL |
Each for $408.50
|
|
|||||
PIMA537189
![]() |
Invitrogen™
MA537189 |
200 μL |
Each for $774.00
|
|
|||||
Description
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.Specifications
| Phospho-MARCKS (Ser167, Ser170) | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide | |
| 80K protein; 80K-L; 80K-L protein; FLJ14368; FLJ90045; KINC; Macs; MARCKS; MARCS; myristoylated alanine rich protein kinase C substrate; myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein; protein kinase C substrate, 80 kDa protein, light chain | |
| MARCKS | |
| IgG κ | |
| Protein A/G | |
| Antibody |
| MARCKSS167170-C9 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P26645, P29966, P30009 | |
| 17118, 25603, 4082 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser167/170 of human phospho MARCKS. | |
| Primary | |
| -20°C | |
| MARCKS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title