Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MARK Pan (Thr215) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105814
Description
Antibody detects endogenous levels of MARK1/2/3/4 only when phosphorylated at Thr215.
MARK1 (MAP/microtubule affinity-regulating kinase 1) belongs to the MARK family of serine/threonine kinases. MARK family protein kinases phosphorylate microtubule-associated proteins and may play a role in cytoskeletal stability. MARK2 refers to MAP/microtubule affinity-regulating kinase 2 isoform a [Homo sapiens]. EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1 called MARK2. MARK3 was originally identified as a marker that was induced by treatment with DNA damaging agents, and loss of MARK3 was found with carcinogenesis in the pancreas. MARK4 contains an N terminal serine/threonine kinase domain, a central ubiquitin associated domain, and a C terminal KA1 associated kinase domain. RT PCR analysis detects upregulated expression of the gene for MARK4 in nearly all clinical hepatocellular carcinoma cells.
Specifications
Phospho-MARK Pan (Thr215) | |
Polyclonal | |
Unconjugated | |
MARK3 | |
1600015G02Rik; 2410090P21Rik; A430080F22Rik; AW491150; B930025N23Rik; C79806; Cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk; EMK1; EMK-1; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; Emk3; ETK1; ETK-1; KIAA1477; Kiaa1860; KP78; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity regulating kinase 2; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MAP/microtubule affinity-regulating kinase 3; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; MARK; MARK1; mark1 {ECO:0000312; MARK2; mark2 {ECO:0000312; MARK3; MARK4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 1; microtubule affinity regulating kinase 2; microtubule affinity regulating kinase 3; microtubule affinity regulating kinase 4; mKIAA1477; mPar-1c; MPK10; MPK-10; Par-1; PAR1 homolog; PAR1 homolog b; PAR1 homolog c; PAR1A; Par-1a; Par1b; Par-1b; Par1c; Par-1c; PAR-1D; protein kinase STK10; RGD:619882}; RGD:708483}; Ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; serine/threonine-protein kinase MARK1; Serine/threonine-protein kinase MARK2; serine/threonine-protein kinase p78; testicular tissue protein Li 117 | |
Rabbit | |
Sequential chromatography | |
RUO | |
117016, 13728, 170577, 17169, 2011, 226778, 232944, 4139, 4140, 57787, 60328, 680407 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08678, O08679, P27448, Q03141, Q05512, Q7KZI7, Q8CIP4, Q8VHF0, Q8VHJ5, Q96L34, Q9P0L2 | |
MARK1, MARK2, MARK3, MARK4 | |
A synthesized peptide derived from human MARK1(Accession Q9P0L2), corresponding to amino acid residues around phosphorylated Thr215. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction