Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MARK4 (Ser423) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986124
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6124 Supplier Bioss Supplier No. BS4660R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the microtubule affinity-regulating kinase family. These protein kinases phosphorylate microtubule-associated proteins and regulate the transition between stable and dynamic microtubules. The encoded protein is associated with the centrosome throughout mitosis and may be involved in cell cycle control. Expression of this gene is a potential marker for cancer, and the encoded protein may also play a role in Alzheimer′s disease. Pseudogenes of this gene are located on both the short and long arm of chromosome 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

Specifications

Antigen Phospho-MARK4 (Ser423)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mark4
Gene Accession No. Q96L34
Gene Alias 2410090P21Rik; C79806; FLJ90097; Kiaa1860; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; Mark4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 4; Nbla00650; PAR-1D
Gene Symbols Mark4
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MARK4 around the phosphorylation site of Ser423.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57787
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.