Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MBP (T98) Mouse anti-Human, Mouse, Rat, Bovine, Clone: 399126, R&D Systems™

Catalog No. MAB4228SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4228SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4228SP Supplier R&D Systems Supplier No. MAB4228SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phospho-MBP (T98) Monoclonal antibody MAB4228SP specifically detects Phospho-MBP (T98) in Bovine, Human, Mouse, Rat samples. It is validated for Western Blotting

Specifications

Antigen MBP (p thr98)
Applications Western Blot
Classification Monoclonal
Clone 399126
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias Hmbpr, MGC99675, mld, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi
Gene Symbols MBP
Host Species Mouse
Immunogen Phosphopeptide containing human MBP T98 site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4155
Test Specificity Detects in vitro-phosphorylated MBP (T98) in Western blots. Based on sequence homology of the immunogen, this antibody should react with phosphorylated MBP from most common vertebrate species, including human, mouse, rat, and bovine.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.