Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MCM2 (Ser53) Recombinant Rabbit Monoclonal Antibody (BLR108H)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | Phospho-MCM2 (Ser53) |
---|---|
Clone | BLR108H |
Concentration | 0.25 mg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544407
|
Invitrogen™
MA544407 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544408
|
Invitrogen™
MA544408 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
Phospho-MCM2 (Ser53) | |
0.25 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
P49736, P97310 | |
17216, 4171 | |
Peptide surrounding phosphorylated serine 53 of Human DNA replication licensing factor MCM2. | |
Antibody |
BLR108H | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
MCM2 | |
AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; Kiaa0030; MCM DNA helicase complex subunit MCM2; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2; Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28; YBL023C; YBL0438 | |
MCM2 | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title