Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MCT1 (Ser213) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105319 Supplier Invitrogen™ Supplier No. PA5105319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of SLC16A1 only when phosphorylated at Ser213.

SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MCT1 (Ser213)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC16A1
Gene Accession No. P53985, P53986, P53987
Gene Alias AL022710; FLJ36745; HHF7; MCT; MCT 1; Mct1; MCT1D; MGC44475; monocarboxylate transporter; Monocarboxylate transporter 1; RATMCT1; RNMCT1; SLC16A1; SLC16A1 protein; solute carrier 16 (monocarboxylic acid transporter), member 1; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Symbols SLC16A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC16A1(Accession P53985), corresponding to amino acid residues around phosphorylated Ser213.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20501, 25027, 6566
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.