Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MDMX (Ser367) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105383
Description
Antibody detects endogenous levels of MDM4 only when phosphorylated at Ser367.
The MDM2 protein is the primary regulator of p53 protein stability. MDMX is an MDM2-related protein that inhibits MDM2-mediated degradation of p53 via distinct associations with MDM2. The gene that encodes MDMX (also designated MDM4) is a target for amplification in malignant gliomas. ARF interacts with MDMX to sequester MDMX within the nucleolus. This sequestration of MDMX by ARF results in an increase in p53 transactivation. In addition, expression of MDMX can reverse MDM2-targeted degradation of p53 while maintaining suppression of p53 transactivation. Like MDM2, MDMX also binds p73 and stabilizes the level of p73. Therefore, in striking contrast to p53, the half-life of p73 is increased by binding to MDM2.
Specifications
Phospho-MDMX (Ser367) | |
Polyclonal | |
Unconjugated | |
MDM4 | |
4933417N07Rik; AA414968; AL023055; AU018793; AU021806; C85810; DKFZp781B1423; double minute 4 homolog; double minute 4 protein; double minute 4, human homolog of; p53-binding protein; HDMX; human homolog of; Mdm2-like p53-binding protein; MDM4; Mdm4 p53 binding protein homolog; MDM4 protein variant G; MDM4 protein variant Y; MDM4, p53 regulator; MDM4-related protein 1; Mdmx; MGC132766; MRP1; p53-binding protein Mdm4; Protein Mdm4; protein Mdmx; RP11-430C7.1; transformed mouse 3T3 cell double minute 4 | |
Rabbit | |
Sequential Chromatography | |
RUO | |
17248, 304798, 4194 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15151, O35618, Q5XIN1 | |
MDM4 | |
A synthesized peptide derived from human MDM4(Accession O15151), corresponding to amino acid residues around phosphorylated Ser367. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction