Learn More
Invitrogen™ Phospho-MECP2 (Ser80) Polyclonal Antibody

Description
This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-MECP2 (Ser80) |
| Applications | Dot Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Mecp2 |
| Gene Accession No. | P51608 |
| Gene Alias | 1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10 |
| Gene Symbols | Mecp2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.