Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MEF2D (Ser444) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986610
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6610 Supplier Bioss Supplier No. BS5502R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the myocyte-specific enhancer factor 2 (MEF2) family of transcription factors. Members of this family are involved in control of muscle and neuronal cell differentiation and development, and are regulated by class II histone deacetylases. Fusions of the encoded protein with Deleted in Azoospermia-Associated Protein 1 (DAZAP1) due to a translocation have been found in an acute lymphoblastic leukemia cell line, suggesting a role in leukemogenesis. The encoded protein may also be involved in Parkinson disease and myotonic dystrophy. Alternative splicing results in multiple transcript variants.

Specifications

Antigen Phospho-MEF2D (Ser444)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MEF2D
Gene Accession No. Q14814
Gene Alias BOS_2847; C80750; DKFZp686I1536; LOC100340589; MADS box transcription enhancer factor 2, polypeptide D; MEF2D; MEF2D/DAZAP1 fusion; myocyte enhancer factor 2D; myocyte enhancer factor 2D-like; Myocyte-specific enhancer factor 2D
Gene Symbols MEF2D
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human MEF2D around the phosphorylation site of Ser444.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4209
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.