Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MEK1/MEK2 (Ser221) Recombinant Rabbit Monoclonal Antibody (MEK12S221-D3), FITC, Invitrogen™

Catalog No. PIMA537330
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537330 10 Tests
PIMA537078 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537330 Supplier Invitrogen™ Supplier No. MA537330
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MEK 1 (MAP Kinase Kinase, also known as MKK) is an integral component of the MAP kinase cascade that regulates cell growth and differentiation (Ahn, 1993; Chong et al., 2003). In brain, this pathway also plays a key role in synaptic (Adams and Sweatt, 2002). Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase (Kyriakis et al., 1991; Seger et al., 1991; Crews et al., 1992).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MEK1/MEK2 (Ser221)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone MEK12S221-D3
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MAP2K2
Gene Accession No. P31938, P36507, Q02750, Q63932
Gene Alias AA589381; CFC3; CFC4; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 2; EC 2.7.12.2; ERK activator kinase 1; ERK activator kinase 2; hypothetical protein; hypothetical protein LOC406728; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP kinase/Erk kinase 1; Map2k1; Map2k2; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK1; MAPKK2; MEK 1; MEK 2; Mek1; MEK-1; Mek2; MEK-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; mitogen-activated protein kinase kinase type 2; MK2; MKK1; MKK2; MP2K1; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen activated, kinase 2, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); si:ch211-242m18.2; wu:fj56a12; wu:fj61b01; zgc:56557; zgc:56557 protein
Gene Symbols MAP2K1, MAP2K2
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser221 of human phospho MEK1/2.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26395, 26396, 5604, 5605
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.