Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MEK3/MEK6 (Ser189, Ser207) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517887
Description
It is not recommended to aliquot this antibody.
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases, act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
Specifications
Phospho-MEK3/MEK6 (Ser189, Ser207) | |
Polyclonal | |
Unconjugated | |
MAP2K3 | |
AW212142; dual specificity mitogen-activated protein kinase kinase 3; Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 3; MAP kinase kinase 6; Map2k3; Map2k6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 3; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 3; mitogen-activated protein kinase kinase 6; MKK3; MKK6; mMKK3b; OTTHUMP00000166044; Prkmk3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 3; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK2; SKK3; Stress-activated protein kinase kinase 2; stress-activated protein kinase kinase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114495, 26397, 26399, 303200, 5606, 5608 | |
-20°C | |
Liquid |
Western Blot | |
40 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
O09110, P46734, P52564, P70236 | |
MAP2K3, MAP2K6 | |
Synthetic phosphopeptide corresponding to residues around pSer189/207 of human MKK3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction