Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MEK3/MEK6 (Ser189, Ser207) Recombinant Rabbit Monoclonal Antibody (MKK3S189MKK6S207-D3), PE, Invitrogen™
GREENER_CHOICE

Catalog No. PIMA537055
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537055 100 Tests
PIMA537316 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537055 Supplier Invitrogen™ Supplier No. MA537055
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases, act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MEK3/MEK6 (Ser189, Ser207)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone MKK3S189MKK6S207-D3
Conjugate PE
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene MAP2K3
Gene Accession No. O09110, P46734, P52564, P70236
Gene Alias AW212142; dual specificity mitogen-activated protein kinase kinase 3; Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 3; MAP kinase kinase 6; Map2k3; Map2k6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 3; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 3; mitogen-activated protein kinase kinase 6; MKK3; MKK6; mMKK3b; OTTHUMP00000166044; Prkmk3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 3; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK2; SKK3; Stress-activated protein kinase kinase 2; stress-activated protein kinase kinase 3
Gene Symbols MAP2K3, MAP2K6
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser189 of human phospho MKK3 and Ser207 of human phospho MKK6.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26397, 26399, 5606, 5608
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.