Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MEK7 (Ser271) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114604 Supplier Invitrogen™ Supplier No. PA5114604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MKK7 only when phosphorylated at Ser271.

The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase family. This kinase specifically activates MAPK8/JNK1 and MAPK9/JNK2, and this kinase itself is phosphorylated and activated by MAP kinase kinases including MAP3K1/MEKK1, MAP3K2/MEKK2, MAP3K3/MEKK5, and MAP4K2/GCK. This kinase is involved in the signal transduction mediating the cell responses to proinflammatory cytokines, and environmental stresses. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found, but only one transcript variant has been supported and defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MEK7 (Ser271)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MAP2K7
Gene Accession No. O14733, Q4KSH7, Q8CE90
Gene Alias 2; 5930412N11Rik; c-Jun N-terminal kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 7; EMBL:AAX61178.1}; JN; JNK kinase 2; JNK-activating kinase 2; JNKK; JNKK 2; Jnkk2; MAP kinase kinase 7; MAP2K4; map2k7; map2k7 {ECO:0000312; MAPK/ERK kinase 4; MAPK/ERK kinase 7; MAPKK 4; MAPKK 7; MAPKK7; MEK; MEK 4; MEK 7; Mek7; mitogen activated protein kinase kinase 7; mitogen-activated protein kinase kinase 7; Mkk7; PRKMK7; protein kinase, mitogen activated, kinase 7; SAPK kinase 1; SAPK kinase 4; SAPK/ERK kinase 1; SAPKK1; SAPKK-1; SAPKK4; SAPKK-4; SEK1; sek2; SKK4; Stress-activated protein kinase kinase 4
Gene Symbols MAP2K7
Host Species Rabbit
Immunogen A synthesized peptide derived from human MKK7 around the phosphorylation site of Ser271.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26400, 363855, 5609
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.