Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MER/SKY (Tyr749, Tyr681) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA537808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537808 Supplier Thermo Scientific Supplier No. PA537808

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is NIH-3T3 cells; suggested positive control for IHC is human brain tissue.

Phospho-MER/SKY (Tyr749, Tyr681) Polyclonal antibody specifically detects Phospho-MER/SKY (Tyr749, Tyr681) in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MER/SKY (Tyr749, Tyr681)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MERTK
Gene Accession No. P55144, Q06418, Q12866, Q60805
Gene Alias AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; c-Eyk; c-mer; c-mer proto-oncogene tyrosine kinase; Dtk; EC 2.7.10.2; Etk-2; etk2/tyro3; Eyk; kinase Syk; KSYK; Mer; MER proto-oncogene, tyrosine kinase; MER receptor tyrosine kinase; MERTK; nmf12; Nyk; Proto-oncogene c-Mer; proto-oncogene tyrosine-protein kinase MER; rdy; Receptor tyrosine kinase MerTK; Receptor tyrosine tinase gene probably the gene for Rdy; Rek; retinal dystrophy; RP38; RSE; Sky; sMER; sMERTK; soluble MER; soluble MERTK; STK kinase; SYK; Tif; TK19-2; Tyro 12; Tyro12; Tyro3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase Mer; Tyrosine-protein kinase receptor TYRO3; tyrosine-protein kinase RSE; Tyrosine-protein kinase SKY; tyrosine-protein kinase TIF
Gene Symbols MERTK, TYRO3
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 749/681(K-I-Y(p)-S-G) derived from Human MER/SKY.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10461, 17289, 22174, 7301
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.