Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MKK7 (Ser271, Thr275) Recombinant Rabbit Monoclonal Antibody (MKK7S271T275-R4F9)

Catalog No. PIMA528042
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528042 200 μL
PIMA537005 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA528042 Supplier Invitrogen™ Supplier No. MA528042
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase family. This kinase specifically activates MAPK8/JNK1 and MAPK9/JNK2, and this kinase itself is phosphorylated and activated by MAP kinase kinases including MAP3K1/MEKK1, MAP3K2/MEKK2, MAP3K3/MEKK5, and MAP4K2/GCK. This kinase is involved in the signal transduction mediating the cell responses to proinflammatory cytokines, and environmental stresses. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found, but only one transcript variant has been supported and defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MKK7 (Ser271, Thr275)
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone MKK7S271T275-R4F9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP2K7
Gene Accession No. O14733, Q4KSH7, Q8CE90
Gene Alias 2; 5930412N11Rik; c-Jun N-terminal kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 7; EMBL:AAX61178.1}; JN; JNK kinase 2; JNK-activating kinase 2; JNKK; JNKK 2; Jnkk2; MAP kinase kinase 7; MAP2K4; map2k7; map2k7 {ECO:0000312; MAPK/ERK kinase 4; MAPK/ERK kinase 7; MAPKK 4; MAPKK 7; MAPKK7; MEK; MEK 4; MEK 7; Mek7; mitogen activated protein kinase kinase 7; mitogen-activated protein kinase kinase 7; Mkk7; PRKMK7; protein kinase, mitogen activated, kinase 7; SAPK kinase 1; SAPK kinase 4; SAPK/ERK kinase 1; SAPKK1; SAPKK-1; SAPKK4; SAPKK-4; SEK1; sek2; SKK4; Stress-activated protein kinase kinase 4
Gene Symbols MAP2K7
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Ser271/Thr275 of human phospho MKK7.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26400, 363855, 5609
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.