Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MKP3 (Ser197) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106011 Supplier Invitrogen™ Supplier No. PA5106011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of DUSP6 only when phosphorylated at Ser197.

DUSP6 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP)kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associatedwith cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK2, is expressed in a variety of tissues with the highest levels in heart and pancreas, and unlike most other members of this family, is localized in the cytoplasm. Twotranscript variants encoding different isoforms have been found for the DUSP6 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MKP3 (Ser197)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DUSP6
Gene Accession No. Q16828, Q64346, Q9DBB1
Gene Alias 1300019I03Rik; dual specificity phosphatase 6; dual specificity protein phosphatase 6; Dual specificity protein phosphatase PYST1; Dusp6; DUSP6-ALT; HH19; MAP kinase phosphatase 3; mitogen-activated protein kinase phosphatase 3; Mkp3; MKP-3; PYST1; serine/threonine specific protein phosphatase
Gene Symbols DUSP6
Host Species Rabbit
Immunogen A synthesized peptide derived from human DUSP6(Accession Q16828), corresponding to amino acid residues around phosphorylated Ser197.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116663, 1848, 67603
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.