Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MLF1 (Thr78)Rabbit anti-Bovine, Canine, Chimpanzee, Human, Polyclonal, Rockland™
SDP

Catalog No. 501991875
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1875 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1875 Supplier Rockland Immunochemicals Supplier No. 600401930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human MLF1IP protein phosphorylated at Thr78. A BLAST analysis was used to suggest cross-reactivity with MLF1IP protein from human, dog, bovine and chimpanzee based on 100% homology with the immunizing sequence. Expect partial reactivity with homologues from rat and mouse (90% homology). Reactivity against homologues from other sources is not known.

The function of this protein remains unknown.

Specifications

Antigen Phospho-MLF1 (Thr78)
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Cenpu
Gene Accession No. Q2KIW5, Q71F23
Gene Alias 1700029A22Rik; CENP50; CENP-50; Cenpu; CENP-U; CENPU50; Centromere protein of 50 kDa; Centromere protein U; ICEN24; Interphase centromere complex protein 24; KLIP1; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; MLF1 interacting protein; MLF1-interacting protein; Mlf1ip; myeloid leukemia factor 1 interacting protein; PBIP1; Polo-box-interacting protein 1
Gene Symbols Cenpu
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 461630, 607799, 617866, 79682
Target Species Bovine, Canine, Chimpanzee, Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.