Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MLK3 (Ser674) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540303
Description
This antibody is predicted to react with human and rat based on sequence homology.
MAP3K11 (MLK3) is a serine/threonine kinase that activates both the p38 MAP kinase and c-Jun kinase pathways. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42.
Specifications
Phospho-MLK3 (Ser674) | |
Polyclonal | |
Unconjugated | |
MAP3K11 | |
2610017K16Rik; MAP3K11; MEKK11; MGC17114; mitogen activated protein kinase kinase kinase 11; mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; Mlk3; MLK-3; protein-tyrosine kinase PTK1; PTK1; RHOE; SH3 domain-containing proline-rich kinase; SPRK; Src-homology 3 domain-containing proline-rich kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26403, 309168, 4296 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q16584, Q66HA1, Q80XI6 | |
MAP3K11 | |
Synthesized peptide derived from human MLK3 around the phosphorylation site of S674. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction