Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-MLKL (Ser345) Mouse anti-Mouse, Clone: 7C6.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1158
Description
Detect phospho MLKL Ser345 using this mouse monoclonal Anti-phospho-MLKL (Ser345), clone 7C6.1 Immunocytochemistry, and Western Blotting.
Specifications
phospho-MLKL (Ser345) | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Mlkl | |
KLH-conjugated linear peptide corresponding to 19 amino acids surrounding Ser345 from the C-terminal half of murine Mixed lineage kinase domain-like protein (MLKL). | |
100 μL | |
Apoptosis and Cancer | |
NP_001297542 | |
Mouse | |
Purified |
Immunocytochemistry, Western Blot, ELISA | |
7C6.1 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 7C6.1 specifically detects MLKL phosphorylalated on Ser345. It targets a sequence of 19 amino acids surrounding Ser345 in the C-terminal region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction