Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MLKL (Ser345) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121360 Supplier Invitrogen™ Supplier No. PA5121360
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa. The target is usually found in the following locations: Cell membrane, Cytoplasm. Immunogen sequence: TQNSIS.

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MLKL (Ser345)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Mlkl
Gene Accession No. Q9D2Y4
Gene Alias 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Gene Symbols Mlkl
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S345 of mouse MLKL (NP_001297542.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74568
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.