Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MLKL (Ser345) Rabbit anti-Human, Clone: 7G4, Bioss
SDP

Catalog No. 501989722
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9722 Supplier Bioss Supplier No. BSM54104R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.

Specifications

Antigen Phospho-MLKL (Ser345)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7G4
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene Mlkl
Gene Accession No. Q8NB16
Gene Alias 9130019I15Rik; hMLKL; mixed lineage kinase domain-like; Mixed lineage kinase domain-like protein; MLKL; rCG51567-like
Gene Symbols Mlkl
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser345 of human MLKL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197259
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.