Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MLKL (Ser358) Recombinant Rabbit Monoclonal Antibody (4A6)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549780
Description
Phospho-MLKL (Ser358) Recombinant Monoclonal Antibody for IHC (P), ELISA
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications
Phospho-MLKL (Ser358) | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NB16 | |
Mlkl | |
A synthesized peptide derived from human Phospho-MLKL (S358). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
4A6 | |
Unconjugated | |
Mlkl | |
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
Rabbit | |
Affinity chromatography | |
RUO | |
197259 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction