Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MLKL (Ser441) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117260
Description
Positive Samples: C6 Immunogen sequence: EPSQR.
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications
Phospho-MLKL (Ser441) | |
Polyclonal | |
Unconjugated | |
Mlkl | |
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
Rabbit | |
Affinity chromatography | |
RUO | |
197259, 690743 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q8NB16 | |
Mlkl | |
A synthetic phosphorylated peptide around S441 of mouse MLKL (NP_0012975421). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction