Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MOK (Tyr161) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564595
Description
Phospho-RAGE (Tyr161) Polyclonal Antibody detects endogenous levels of RAGE only when phosphorylated at Tyr161.
MOK is able to phosphorylate several exogenous substrates and to undergo autophosphorylation. It negatively regulates cilium length in a cAMP and mTORC1 signaling-dependent manner.
Specifications
Phospho-MOK (Tyr161) | |
Polyclonal | |
Unconjugated | |
MOK | |
MAPK/MAK/MRK overlapping kinase; MAPK/MAK/MRK/ overlapping kinase; MOK; MOK protein kinase; Rage; RAGE1; RAGE-1; renal cell carcinoma antigen; renal tumor antigen; renal tumor antigen 1; Serine/threonine kinase 30; soluble RAGE; STK30; T/STK 30 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26448, 362787, 5891 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UQ07, Q9WVS4 | |
MOK | |
A synthesized peptide derived from human MOK (Phospho-Tyr161). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction