Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-MPRIP (Thr542) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105059
Description
Antibody detects endogenous levels of MRIP only when phosphorylated at Thr542.
Targets myosin phosphatase to the actin cytoskeleton. Required for the regulation of the actin cytoskeleton by RhoA and ROCK1. Depletion leads to an increased number of stress fibers in smooth muscle cells through stabilization of actin fibers by phosphorylated myosin. Overexpression of MRIP as well as its F-actin-binding region leads to disassembly of stress fibers in neuronal cells.
Specifications
| Phospho-MPRIP (Thr542) | |
| Polyclonal | |
| Unconjugated | |
| MPRIP | |
| 9530046C02; AA536749; AI647711; C76423; KIAA0864; mKIAA0864; Mprip; Mrip; M-RIP; myosin phosphatase Rho interacting protein; myosin phosphatase Rho-interacting protein; myosin phosphatase-Rho interacting protein; p116 Rho interacting protein; p116Rip; Rho interacting protein 3; rho-interacting protein 3; Rho-interacting protein 3 (p116RIP) (RIP3); RHOIP3; RIP3 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 116504, 23164, 26936 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97434, Q6WCQ1, Q9ERE6 | |
| MPRIP | |
| A synthesized peptide derived from human MPRIP(Accession Q6WCQ1), corresponding to amino acid residues around phosphorylated Thr542. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction