Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MRE11 (Ser264) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538868
Description
MRE11 (double-strand break repair protein MRE11) is a component of the MRN complex. This complex plays a central role in double-strand break repair, DNA recombination, maintece of telomere integrity and meiosis. The complex possesses single-stranded endonuclease activity and double-strand-specific 3′-5′ exonuclease activity which are provided by MRE11. RAD50 is required to bind DNA ends and hold them in close proximity. Mutations in the gene could result in ataxia-telangiectasia-like disorder 1.
Specifications
Phospho-MRE11 (Ser264) | |
Polyclonal | |
Unconjugated | |
MRE11 | |
AT-like disease, DNA recombination and repair protein, MRE11 homolog 1, MRE11 homolog A, double-strand break repair protein MRE11A, endo/exonuclease Mre11, meiotic recombination 11 homolog 1, meiotic recombination 11 homolog A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17535, 4361 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49959, Q61216 | |
MRE11, Mre11a | |
A synthetic phosphopeptide derived from human MRE11 around the phosphorylation site of Ser264 (Y-I-SP-Q-P) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction