Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Mre11 (Ser676) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985283
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5283 Supplier Bioss Supplier No. BS3293R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRE11 (double-strand break repair protein MRE11) is a component of the MRN complex. This complex plays a central role in double-strand break repair, DNA recombination, maintenance of telomere integrity and meiosis. The complex possesses single-stranded endonuclease activity and double-strand-specific 3′-5′ exonuclease activity which are provided by MRE11. RAD50 is required to bind DNA ends and hold them in close proximity. Mutations in the gene could result in ataxia-telangiectasia-like disorder 1.

Specifications

Antigen Phospho-Mre11 (Ser676)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mre11
Gene Accession No. P49959
Gene Alias ATLD; AT-like disease; DNA recombination and repair protein; Double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; Meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); MmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; Mre11a; MRE11B
Gene Symbols Mre11
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from rat Mre11 around the phosphorylation site of Ser676.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4361
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.